Product Details

SNP ID
rs77083413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140631967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGGAGTTCATTGAGCCCTCGTG[C/G]GGGAGGGCAGTTCCAGGGACCAGGA
Phenotype
MIM: 158120
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD14 PubMed Links

Gene Details

Gene
CD14
Gene Name
CD14 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000591.3 1409 Silent Mutation CCC,CCG P339P NP_000582.1
NM_001040021.2 1409 Silent Mutation CCC,CCG P339P NP_001035110.1
NM_001174104.1 1409 Silent Mutation CCC,CCG P339P NP_001167575.1
NM_001174105.1 1409 Silent Mutation CCC,CCG P339P NP_001167576.1
Gene
TMCO6
Gene Name
transmembrane and coiled-coil domains 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300980.1 1409 Intron NP_001287909.1
NM_001300982.1 1409 Intron NP_001287911.1
NM_018502.4 1409 Intron NP_060972.3
XM_005268477.1 1409 Intron XP_005268534.1
XM_011537663.2 1409 Intron XP_011535965.1
XM_011537665.2 1409 Intron XP_011535967.1
XM_011537668.2 1409 Intron XP_011535970.1
XM_017009617.1 1409 Intron XP_016865106.1
XM_017009618.1 1409 Intron XP_016865107.1
XM_017009619.1 1409 Intron XP_016865108.1
XM_017009620.1 1409 Intron XP_016865109.1

View Full Product Details