Product Details

SNP ID
rs79385276
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140705696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGACTTGACATTTGAGGAGGA[C/T]GATGAGATGGCAGCTGTGATGGGCT
Phenotype
MIM: 600783 MIM: 612695
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HARS2 PubMed Links

Gene Details

Gene
HARS2
Gene Name
histidyl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.

Gene
VTRNA1-1
Gene Name
vault RNA 1-1
There are no transcripts associated with this gene.

Gene
ZMAT2
Gene Name
zinc finger matrin-type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144723.2 641 Silent Mutation GAC,GAT D180D NP_653324.1

View Full Product Details