Product Details

SNP ID
rs79034785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:154448492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTGATCCCTCCTTAGGTCACAC[A/G]TTGAGGTGAAATGCTTATGGCAATG
Phenotype
MIM: 610398
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAP30L PubMed Links

Gene Details

Gene
SAP30L
Gene Name
SAP30 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131062.1 Intron NP_001124534.1
NM_001131063.1 Intron NP_001124535.1
NM_024632.5 Intron NP_078908.1
Gene
SAP30L-AS1
Gene Name
SAP30L antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details