Product Details

SNP ID
rs77597690
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:148206857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCTTGGCGTGCCTTGTTATACC[G/T]TTGACATGAAAATTTCTAGACACTA
Phenotype
MIM: 615868
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
SPINK6 PubMed Links

Gene Details

Gene
SPINK6
Gene Name
serine peptidase inhibitor, Kazal type 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195290.1 Intron NP_001182219.1
NM_205841.3 Intron NP_995313.2

View Full Product Details