Product Details

SNP ID
rs75075713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140423674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTTTCATTTAAGAGACCTTCCA[A/G]CTATTAATACCTACTGTATGTATAT
Phenotype
MIM: 610500
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKHD1 PubMed Links

Gene Details

Gene
ANKHD1
Gene Name
ankyrin repeat and KH domain containing 1
There are no transcripts associated with this gene.

Gene
ANKHD1-EIF4EBP3
Gene Name
ANKHD1-EIF4EBP3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020690.5 Intron NP_065741.3

View Full Product Details