Product Details

SNP ID
rs76904460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:39295022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTAGGCAGGGAGAAAGCTGGGGC[A/C]GGGTTTCTAGGGAGTTAGCGAATCT
Phenotype
MIM: 608283
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KIF21A PubMed Links

Gene Details

Gene
KIF21A
Gene Name
kinesin family member 21A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173463.1 Intron NP_001166934.1
NM_001173464.1 Intron NP_001166935.1
NM_001173465.1 Intron NP_001166936.1
NM_017641.3 Intron NP_060111.2
XM_005269007.2 Intron XP_005269064.1
XM_005269008.2 Intron XP_005269065.1
XM_005269009.2 Intron XP_005269066.1
XM_005269010.2 Intron XP_005269067.1
XM_005269011.2 Intron XP_005269068.1
XM_005269012.2 Intron XP_005269069.1
XM_005269013.2 Intron XP_005269070.1
XM_005269014.2 Intron XP_005269071.1
XM_006719493.2 Intron XP_006719556.1
XM_006719494.2 Intron XP_006719557.1
XM_011538556.2 Intron XP_011536858.1
XM_017019607.1 Intron XP_016875096.1
XM_017019608.1 Intron XP_016875097.1
XM_017019609.1 Intron XP_016875098.1
XM_017019610.1 Intron XP_016875099.1
XM_017019611.1 Intron XP_016875100.1

View Full Product Details