Product Details

SNP ID
rs77145008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55954483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGCTTCCTCCCCTTCTCCTCACC[C/T]CCAGGGTTTACCCTCTCTTATTTCT
Phenotype
MIM: 125855 MIM: 155550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGKA PubMed Links

Gene Details

Gene
DGKA
Gene Name
diacylglycerol kinase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001345.4 Intron NP_001336.2
NM_201444.2 Intron NP_958852.1
NM_201445.1 Intron NP_958853.1
NM_201554.1 Intron NP_963848.1
XM_005268688.2 Intron XP_005268745.1
XM_005268689.2 Intron XP_005268746.1
XM_005268690.2 Intron XP_005268747.1
XM_011537991.2 Intron XP_011536293.1
XM_011537993.2 Intron XP_011536295.1
XM_011537995.2 Intron XP_011536297.1
XM_017018900.1 Intron XP_016874389.1
XM_017018901.1 Intron XP_016874390.1
XM_017018902.1 Intron XP_016874391.1
XM_017018903.1 Intron XP_016874392.1
XM_017018904.1 Intron XP_016874393.1
XM_017018905.1 Intron XP_016874394.1
XM_017018906.1 Intron XP_016874395.1
XM_017018907.1 Intron XP_016874396.1
XM_017018908.1 Intron XP_016874397.1
XM_017018909.1 Intron XP_016874398.1
Gene
PMEL
Gene Name
premelanosome protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001200053.1 Intron NP_001186982.1
NM_001200054.1 Intron NP_001186983.1
NM_001320121.1 Intron NP_001307050.1
NM_001320122.1 Intron NP_001307051.1
NM_006928.4 Intron NP_008859.1
XM_006719569.1 Intron XP_006719632.1
XM_011538685.1 Intron XP_011536987.1

View Full Product Details