Product Details

SNP ID
rs79466051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:64193367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGATCAGAATCATAAATCTACT[A/G]ACTCCATATTCATTTGCTAATTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf66 PubMed Links

Gene Details

Gene
C12orf66
Gene Name
chromosome 12 open reading frame 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300940.1 1713 Intron NP_001287869.1
NM_001300941.1 1713 UTR 3 NP_001287870.1
NM_152440.4 1713 UTR 3 NP_689653.3

View Full Product Details