Product Details

SNP ID
rs74955204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:41188817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGCCGCGGGGGCTGCGCTTCGG[A/G]GCTGGGCGGTGGTGAGGTGCCCGCG
Phenotype
MIM: 609730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDZRN4 PubMed Links

Gene Details

Gene
PDZRN4
Gene Name
PDZ domain containing ring finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164595.1 370 Missense Mutation GAG,GGG E121G NP_001158067.1
NM_013377.3 370 Intron NP_037509.3

View Full Product Details