Product Details

SNP ID
rs77407708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:96502632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAAACAGTGTTTGACTGAAGAT[A/G]CCTCCCAAGGGGCTAGGTAACTGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP54 PubMed Links

Gene Details

Gene
CFAP54
Gene Name
cilia and flagella associated protein 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306084.1 Intron NP_001293013.1
XM_011539069.2 Intron XP_011537371.1
XM_011539070.2 Intron XP_011537372.1
XM_011539071.2 Intron XP_011537373.1
XM_011539072.2 Intron XP_011537374.1
XM_011539073.2 Intron XP_011537375.1
XM_011539074.2 Intron XP_011537376.1
XM_011539076.2 Intron XP_011537378.1
XM_011539077.2 Intron XP_011537379.1
XM_011539078.2 Intron XP_011537380.1
XM_011539079.1 Intron XP_011537381.1
XM_011539081.2 Intron XP_011537383.1
XM_011539083.2 Intron XP_011537385.1
XM_011539084.2 Intron XP_011537386.1
XM_011539085.1 Intron XP_011537387.1
XM_017018866.1 Intron XP_016874355.1
XM_017018867.1 Intron XP_016874356.1

View Full Product Details