Product Details

SNP ID
rs78877410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:107317606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAGACAAGTAGATGACACGATG[C/T]TATTTGACACAGGAATGCAGCTAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTBD11 PubMed Links

Gene Details

Gene
BTBD11
Gene Name
BTB domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017523.1 439 Intron NP_001017523.1
NM_001018072.1 439 Intron NP_001018082.1
XM_005268645.3 439 Intron XP_005268702.1
XM_011537909.2 439 Intron XP_011536211.1
XM_011537910.2 439 Intron XP_011536212.1
XM_017018807.1 439 UTR 5 XP_016874296.1

View Full Product Details