Product Details

SNP ID
rs76511847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:62806770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGATTCAAAGGAGCTGAATGT[G/T]TTATGCTTCCAAACAACTGAATGTA
Phenotype
MIM: 611392 MIM: 129010
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADO PubMed Links

Gene Details

Gene
ADO
Gene Name
2-aminoethanethiol dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032804.5 2015 UTR 3 NP_116193.2
Gene
EGR2
Gene Name
early growth response 2
There are no transcripts associated with this gene.

View Full Product Details