Product Details

SNP ID
rs80200823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49609164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTCAGCCTGTCAGCTGAGCAC[C/G]GGCGCCTCAAGGGGTGCGGCCCTCT
Phenotype
MIM: 118490 MIM: 600336
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHAT PubMed Links

Gene Details

Gene
CHAT
Gene Name
choline O-acetyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142929.1 70 Intron NP_001136401.1
NM_001142933.1 70 Intron NP_001136405.1
NM_001142934.1 70 Intron NP_001136406.1
NM_020549.4 70 Intron NP_065574.3
NM_020984.3 70 UTR 5 NP_066264.3
NM_020985.3 70 Intron NP_066265.3
NM_020986.3 70 Intron NP_066266.3
Gene
SLC18A3
Gene Name
solute carrier family 18 member A3
There are no transcripts associated with this gene.

View Full Product Details