Product Details

SNP ID
rs77201605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:74122333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATCCTTAGCATAAGGGAAAGAC[A/G]GTTATAAGCTGAGAAGATTGAAAGA
Phenotype
MIM: 610366 MIM: 193065
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP3M1 PubMed Links

Gene Details

Gene
AP3M1
Gene Name
adaptor related protein complex 3 mu 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320263.1 3068 UTR 3 NP_001307192.1
NM_001320264.1 3068 UTR 3 NP_001307193.1
NM_001320265.1 3068 UTR 3 NP_001307194.1
NM_012095.5 3068 UTR 3 NP_036227.1
NM_207012.3 3068 UTR 3 NP_996895.1
Gene
VCL
Gene Name
vinculin
There are no transcripts associated with this gene.

View Full Product Details