Product Details

SNP ID
rs78703311
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:77976030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCATTTTTTTTCCACATCACTGA[C/T]ACCCCAAGAGAAGTTAATAGGTCTA
Phenotype
MIM: 614258
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
POLR3A PubMed Links

Gene Details

Gene
POLR3A
Gene Name
RNA polymerase III subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007055.3 5754 UTR 3 NP_008986.2

View Full Product Details