Product Details

SNP ID
rs75460698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84340342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTGACAGGTTGCTCTCTTGCCCC[C/T]ACTTATAGACACTCAATAAATTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCSER2 PubMed Links

Gene Details

Gene
CCSER2
Gene Name
coiled-coil serine rich protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284240.1 Intron NP_001271169.1
NM_001284241.1 Intron NP_001271170.1
NM_001284242.1 Intron NP_001271171.1
NM_001284243.1 Intron NP_001271172.1
NM_018999.3 Intron NP_061872.2
XM_005269905.3 Intron XP_005269962.1
XM_005269906.3 Intron XP_005269963.1
XM_005269908.3 Intron XP_005269965.1
XM_006717894.1 Intron XP_006717957.1
XM_006717895.1 Intron XP_006717958.1
XM_011539870.2 Intron XP_011538172.1
XM_011539871.2 Intron XP_011538173.1
XM_011539872.2 Intron XP_011538174.1
XM_011539873.1 Intron XP_011538175.1
XM_017016340.1 Intron XP_016871829.1
XM_017016341.1 Intron XP_016871830.1
XM_017016342.1 Intron XP_016871831.1
XM_017016343.1 Intron XP_016871832.1

View Full Product Details