Product Details

SNP ID
rs78286608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96366627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTGTAAAAAATATAACCTTTCA[C/T]CTTTTACATATACTTTGTTTAAAAT
Phenotype
MIM: 606743
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OPALIN PubMed Links

Gene Details

Gene
OPALIN
Gene Name
oligodendrocytic myelin paranodal and inner loop protein
There are no transcripts associated with this gene.

Gene
TLL2
Gene Name
tolloid like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012465.3 4750 UTR 3 NP_036597.1

View Full Product Details