Product Details

SNP ID
rs76145370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103398247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTTTGAAGCAGAGACCTTTGGA[A/G]AGATGAAGTATGGATTATTCCGGGC
Phenotype
MIM: 612333 MIM: 601787 MIM: 615204
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR1307 PubMed Links

Gene Details

Gene
MIR1307
Gene Name
microRNA 1307
There are no transcripts associated with this gene.

Gene
PDCD11
Gene Name
programmed cell death 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014976.1 Intron NP_055791.1
XM_005269647.3 Intron XP_005269704.1
XM_011539538.2 Intron XP_011537840.1
XM_011539539.2 Intron XP_011537841.1
XM_011539540.1 Intron XP_011537842.1
Gene
TAF5
Gene Name
TATA-box binding protein associated factor 5
There are no transcripts associated with this gene.

Gene
USMG5
Gene Name
up-regulated during skeletal muscle growth 5 homolog (mouse)
There are no transcripts associated with this gene.

View Full Product Details