Product Details

SNP ID
rs78125145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103403268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAAGCTGAATGAGCAGGTGACA[A/C]AAGAACAACCTCTGAAGGTAAGGGA
Phenotype
MIM: 612333 MIM: 615204
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR1307 PubMed Links

Gene Details

Gene
MIR1307
Gene Name
microRNA 1307
There are no transcripts associated with this gene.

Gene
PDCD11
Gene Name
programmed cell death 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014976.1 512 Missense Mutation AAA,CAA K129Q NP_055791.1
XM_005269647.3 512 Missense Mutation AAA,CAA K129Q XP_005269704.1
XM_011539538.2 512 Missense Mutation AAA,CAA K129Q XP_011537840.1
XM_011539539.2 512 Missense Mutation AAA,CAA K129Q XP_011537841.1
XM_011539540.1 512 Missense Mutation AAA,CAA K46Q XP_011537842.1
Gene
USMG5
Gene Name
up-regulated during skeletal muscle growth 5 homolog (mouse)
There are no transcripts associated with this gene.

View Full Product Details