Product Details

SNP ID
rs76268637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:112454897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTCTGATCTTATTTAATGGCTT[A/G]AGAAAGGGATAGTAATAATACCTCA
Phenotype
MIM: 614316
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VTI1A PubMed Links

Gene Details

Gene
VTI1A
Gene Name
vesicle transport through interaction with t-SNAREs 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318203.1 Intron NP_001305132.1
NM_001318205.1 Intron NP_001305134.1
NM_145206.3 Intron NP_660207.2
XM_005269544.4 Intron XP_005269601.1
XM_005269545.4 Intron XP_005269602.1
XM_005269546.3 Intron XP_005269603.1
XM_005269547.4 Intron XP_005269604.1
XM_006717637.2 Intron XP_006717700.1
XM_011539328.2 Intron XP_011537630.1
XM_011539330.2 Intron XP_011537632.1
XM_011539331.2 Intron XP_011537633.1
XM_011539333.2 Intron XP_011537635.1
XM_017015745.1 Intron XP_016871234.1
XM_017015746.1 Intron XP_016871235.1
XM_017015747.1 Intron XP_016871236.1
XM_017015748.1 Intron XP_016871237.1
Gene
ZDHHC6
Gene Name
zinc finger DHHC-type containing 6
There are no transcripts associated with this gene.

View Full Product Details