Product Details

SNP ID
rs75483954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:113681032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGGCAAGAAGTGAGTCACAAAT[C/T]GTATGATGTCACTTTTCAGGGGAAG
Phenotype
MIM: 601761
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASP7 PubMed Links

Gene Details

Gene
CASP7
Gene Name
caspase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001227.4 Intron NP_001218.1
NM_001267056.1 Intron NP_001253985.1
NM_001267057.1 Intron NP_001253986.1
NM_001267058.1 Intron NP_001253987.1
NM_001320911.1 Intron NP_001307840.1
NM_033338.5 Intron NP_203124.1
NM_033339.4 Intron NP_203125.1
NM_033340.3 Intron NP_203126.1
XM_006718017.3 Intron XP_006718080.1
XM_011540260.1 Intron XP_011538562.1
XM_017016763.1 Intron XP_016872252.1
XM_017016764.1 Intron XP_016872253.1

View Full Product Details