Product Details

SNP ID
rs74581721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:114122942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTCCAAGTAGATGGTAGCACAA[A/G]TATTAATACTGGAGTGTGCATTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC186 PubMed Links

Gene Details

Gene
CCDC186
Gene Name
coiled-coil domain containing 186
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321829.1 4508 UTR 3 NP_001308758.1
NM_018017.3 4508 UTR 3 NP_060487.2
NM_153249.1 4508 Intron NP_694981.1
XM_011539915.2 4508 UTR 3 XP_011538217.1
XM_011539916.2 4508 Intron XP_011538218.1

View Full Product Details