Product Details

SNP ID
rs76660677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:124007702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAAGAGCACGCGCTCCACAGGC[A/G]TCACTCTTATGGGTCCATCTTTAAT
Phenotype
MIM: 608277
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHST15 PubMed Links

Gene Details

Gene
CHST15
Gene Name
carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270764.1 4698 UTR 3 NP_001257693.1
NM_001270765.1 4698 Intron NP_001257694.1
NM_014863.3 4698 Intron NP_055678.2
NM_015892.4 4698 UTR 3 NP_056976.2
XM_005269891.3 4698 UTR 3 XP_005269948.1
XM_005269892.4 4698 Intron XP_005269949.1
XM_005269893.3 4698 Intron XP_005269950.1
XM_005269894.4 4698 Intron XP_005269951.1
XM_006717891.3 4698 UTR 3 XP_006717954.1
XM_011539857.1 4698 Intron XP_011538159.1
XM_017016319.1 4698 UTR 3 XP_016871808.1
XM_017016320.1 4698 Intron XP_016871809.1
XM_017016321.1 4698 Intron XP_016871810.1

View Full Product Details