Product Details
- SNP ID
-
rs76628300
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
17
- Location
-
Chr.1:42747259 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCACCAGCTGCTCTCACCCGCTC[C/G]AGCTGCTCTCACCCGCTCGCTGTGT
- Phenotype
-
MIM: 610036
MIM: 610339
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN19
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs67014447] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLDN19
- Gene Name
- claudin 19
There are no transcripts associated with this gene.
- Gene
- P3H1
- Gene Name
- prolyl 3-hydroxylase 1
View Full Product Details