Product Details

SNP ID
rs77932721
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:45344272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAATAGGTTAGCTAAGAGCCAT[G/T]ACCACCACGCTGCCTTGCTGTCCCC
Phenotype
MIM: 604933 MIM: 604746 MIM: 613931
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MUTYH PubMed Links

Gene Details

Gene
MUTYH
Gene Name
mutY DNA glycosylase
There are no transcripts associated with this gene.

Gene
TESK2
Gene Name
testis-specific kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320800.1 3015 UTR 3 NP_001307729.1
NM_007170.2 3015 UTR 3 NP_009101.2
XM_005270355.3 3015 UTR 3 XP_005270412.1
XM_006710287.2 3015 UTR 3 XP_006710350.1
XM_011540497.1 3015 UTR 3 XP_011538799.1
XM_011540500.2 3015 Intron XP_011538802.1
XM_017000047.1 3015 UTR 3 XP_016855536.1
XM_017000048.1 3015 UTR 3 XP_016855537.1
XM_017000049.1 3015 UTR 3 XP_016855538.1
Gene
TOE1
Gene Name
target of EGR1, member 1 (nuclear)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025077.3 3015 Intron NP_079353.3
XM_005270412.3 3015 Intron XP_005270469.1
XM_005270413.4 3015 Intron XP_005270470.1
XM_011540569.2 3015 Intron XP_011538871.1

View Full Product Details