Product Details

SNP ID
rs79193797
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:53853646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATACCACGAAGGTGAGGCTAAG[G/T]CAAGGGCTAACTGCCCAAGTCTACT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
YIPF1 PubMed Links

Gene Details

Gene
YIPF1
Gene Name
Yip1 domain family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018982.4 Intron NP_061855.1
XM_017001499.1 Intron XP_016856988.1
XM_017001500.1 Intron XP_016856989.1
XM_017001501.1 Intron XP_016856990.1

View Full Product Details