Product Details

SNP ID
rs79867114
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:54175965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAATTTAAATGAAAAAACCATC[C/T]TACAAGCCCACAAAAAAACAAAATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYB5RL PubMed Links

Gene Details

Gene
CYB5RL
Gene Name
cytochrome b5 reductase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031672.2 Intron NP_001026842.2

View Full Product Details