Product Details

SNP ID
rs79913813
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:62455259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACCATAGCCATGATTCTCAAGC[A/G]TTAACAATCTACATTTGATATTTTC
Phenotype
MIM: 615730 MIM: 603478
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOCK7 PubMed Links

Gene Details

Gene
DOCK7
Gene Name
dedicator of cytokinesis 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271999.1 6573 UTR 3 NP_001258928.1
NM_001272000.1 6573 UTR 3 NP_001258929.1
NM_001272001.1 6573 UTR 3 NP_001258930.1
NM_001272002.1 6573 Intron NP_001258931.1
NM_033407.3 6573 UTR 3 NP_212132.2
XM_005271292.2 6573 UTR 3 XP_005271349.1
XM_011542326.2 6573 UTR 3 XP_011540628.1
XM_011542327.2 6573 UTR 3 XP_011540629.1
XM_011542328.2 6573 UTR 3 XP_011540630.1
XM_011542330.2 6573 Intron XP_011540632.1
XM_017002639.1 6573 UTR 3 XP_016858128.1
XM_017002640.1 6573 Intron XP_016858129.1
Gene
USP1
Gene Name
ubiquitin specific peptidase 1
There are no transcripts associated with this gene.

View Full Product Details