Product Details

SNP ID
rs74225876
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:77991410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCAATACTTGAAATGCTTTCAA[G/T]GTCATTTTTAGACATACGCATGTTC
Phenotype
MIM: 611327
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB4 PubMed Links

Gene Details

Gene
DNAJB4
Gene Name
DnaJ heat shock protein family (Hsp40) member B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317099.1 Intron NP_001304028.1
NM_001317100.1 Intron NP_001304029.1
NM_001317101.1 Intron NP_001304030.1
NM_001317102.1 Intron NP_001304031.1
NM_001317103.1 Intron NP_001304032.1
NM_007034.4 Intron NP_008965.2

View Full Product Details