Product Details

SNP ID
rs77524168
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:34852162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAAAGTGCCCTTGGAAAGGAAAG[A/G]AAAGAATTTAAAAGGGGGCACAATC
Phenotype
MIM: 611413
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLGAP3 PubMed Links

Gene Details

Gene
DLGAP3
Gene Name
DLG associated protein 3
There are no transcripts associated with this gene.

Gene
SMIM12
Gene Name
small integral membrane protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164824.1 3986 UTR 3 NP_001158296.1
NM_001164825.1 3986 UTR 3 NP_001158297.1
NM_001320261.1 3986 UTR 3 NP_001307190.1
NM_138428.5 3986 UTR 3 NP_612437.3
XM_005270403.4 3986 Intron XP_005270460.1
XM_011540553.2 3986 Intron XP_011538855.1
XM_011540554.2 3986 Intron XP_011538856.2
XM_011540555.2 3986 Intron XP_011538857.1
XM_017000166.1 3986 Intron XP_016855655.1

View Full Product Details