Product Details

SNP ID
rs79631927
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:36316033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAACTTCTTCAAATGTTTTATA[C/T]GTATCTATGCAGTGTTCTCTACCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EVA1B PubMed Links

Gene Details

Gene
EVA1B
Gene Name
eva-1 homolog B (C. elegans)
There are no transcripts associated with this gene.

Gene
SH3D21
Gene Name
SH3 domain containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162530.1 Intron NP_001156002.1
NM_024676.4 Intron NP_078952.4
XM_011542153.2 Intron XP_011540455.1
XM_017002340.1 Intron XP_016857829.1
XM_017002341.1 Intron XP_016857830.1
XM_017002342.1 Intron XP_016857831.1
XM_017002343.1 Intron XP_016857832.1
XM_017002344.1 Intron XP_016857833.1
XM_017002345.1 Intron XP_016857834.1
XM_017002346.1 Intron XP_016857835.1

View Full Product Details