Product Details

SNP ID
rs76638061
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:100196184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACAATATAAATTGTAAAGATGAA[C/T]ATGTGCTGGCACAGCTAGGGTTTAC
Phenotype
MIM: 248610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DBT PubMed Links

Gene Details

Gene
DBT
Gene Name
dihydrolipoamide branched chain transacylase E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001918.3 1780 UTR 3 NP_001909.3
XM_005270545.3 1780 UTR 3 XP_005270602.1
XM_017000468.1 1780 UTR 3 XP_016855957.1
XM_017000469.1 1780 UTR 3 XP_016855958.1

View Full Product Details