Product Details

SNP ID
rs76463942
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:108138085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTCACTGACAGCATTCTGCAGC[A/G]CCCAAGTTAGGTTATAGCTCCTTGC
Phenotype
MIM: 608744
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A24 PubMed Links

Gene Details

Gene
SLC25A24
Gene Name
solute carrier family 25 member 24
There are no transcripts associated with this gene.

View Full Product Details