Product Details

SNP ID
rs77292583
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:116910247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCGCTGCTGCTGGCGCTCCTGT[C/G]GTTGGGTGAGTGTGCGCGGGGCTCA
Phenotype
MIM: 601204
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PTGFRN PubMed Links

Gene Details

Gene
PTGFRN
Gene Name
prostaglandin F2 receptor inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020440.3 325 Missense Mutation TCG,TGG S15W NP_065173.2
XM_017001874.1 325 Intron XP_016857363.1

View Full Product Details