Product Details

SNP ID
rs74657953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126426490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTAGCACAGTGCCTGATATACAA[C/T]AGAGCTGCTTCAAATGGTATGTATA
Phenotype
MIM: 607761 MIM: 104240
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIRREL3 PubMed Links

Gene Details

Gene
KIRREL3
Gene Name
kin of IRRE like 3 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161707.1 Intron NP_001155179.1
NM_001301097.1 Intron NP_001288026.1
NM_032531.3 Intron NP_115920.1
XM_011543026.2 Intron XP_011541328.1
XM_011543027.2 Intron XP_011541329.1
XM_011543028.2 Intron XP_011541330.1
XM_011543030.2 Intron XP_011541332.1
XM_011543031.2 Intron XP_011541333.1
XM_011543032.2 Intron XP_011541334.1
XM_011543033.1 Intron XP_011541335.1
XM_017018418.1 Intron XP_016873907.1
XM_017018419.1 Intron XP_016873908.1
XM_017018420.1 Intron XP_016873909.1
Gene
ST3GAL4
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001254757.1 Intron NP_001241686.1
NM_001254758.1 Intron NP_001241687.1
NM_001254759.1 Intron NP_001241688.1
NM_006278.2 Intron NP_006269.1
XM_011542959.1 Intron XP_011541261.1
XM_011542960.2 Intron XP_011541262.1
XM_017018134.1 Intron XP_016873623.1

View Full Product Details