Product Details

SNP ID
rs78585439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:101033319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCACATCTCTGTTTCAACAAGA[A/G]TATGGTATGTTTGTATTAACTCTCC
Phenotype
MIM: 607311
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGR PubMed Links

Gene Details

Gene
PGR
Gene Name
progesterone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000926.4 9342 UTR 3 NP_000917.3
NM_001202474.3 9342 UTR 3 NP_001189403.1
NM_001271161.2 9342 UTR 3 NP_001258090.1
NM_001271162.1 9342 UTR 3 NP_001258091.1
XM_006718858.3 9342 Intron XP_006718921.1
XM_011542869.2 9342 Intron XP_011541171.1

View Full Product Details