Product Details
- SNP ID
-
rs79248469
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:102577426 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTCCTGAAAAGAAGTAAATGTAG[A/C]CTAAAAGAGACAAAGTTGAAATTGG
- Phenotype
-
MIM: 604629
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MMP20
PubMed Links
Gene Details
- Gene
- MMP20
- Gene Name
- matrix metallopeptidase 20
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004771.3 |
1365 |
Missense Mutation |
GGC,GTC |
G451V |
NP_004762.2 |
View Full Product Details