Product Details

SNP ID
rs78429131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:112707687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTTGGTGAGGAAGGTGAAGCAC[G/T]CAGTTGCCTTCTCGGGCCTCGGCGC
Phenotype
MIM: 611731
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
APC PubMed Links

Gene Details

Gene
APC
Gene Name
APC, WNT signaling pathway regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000038.5 183 Intron NP_000029.2
NM_001127510.2 183 Intron NP_001120982.1
NM_001127511.2 183 UTR 5 NP_001120983.2

View Full Product Details