Product Details

SNP ID
rs78257855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:120473229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATCAGAGGTATATTTATGTTTAT[C/T]CAATTATATATAGAATGAAGTATAA
Phenotype
MIM: 615931
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRR16 PubMed Links

Gene Details

Gene
PRR16
Gene Name
proline rich 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300783.1 Intron NP_001287712.1
NM_001308087.1 Intron NP_001295016.1
NM_001308088.1 Intron NP_001295017.1
NM_016644.2 Intron NP_057728.1
XM_011543452.2 Intron XP_011541754.1
XM_011543453.2 Intron XP_011541755.1
XM_011543454.2 Intron XP_011541756.1

View Full Product Details