Product Details

SNP ID
rs75388024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:123099837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACCAGTACTGTGCCCGGCCTGG[C/T]CTACGGCATCTGCGCGGCGCAGAGG
Phenotype
MIM: 616982
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRDM6 PubMed Links

Gene Details

Gene
PRDM6
Gene Name
PR domain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136239.1 1228 Missense Mutation GCC,GTC A259V NP_001129711.1
XM_011543726.2 1228 Missense Mutation GCC,GTC A59V XP_011542028.1

View Full Product Details