Product Details
- SNP ID
-
rs78638498
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:127652340 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCATCAGTTAGCTGGCACCTCGG[A/G]TTTTGAATTCAGTTCCCTGGACTGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTXN3
PubMed Links
Gene Details
- Gene
- CTXN3
- Gene Name
- cortexin 3
View Full Product Details