Product Details
- SNP ID
-
rs79869087
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:138285729 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCCTTCACCAGAAGGGCAATCTGC[C/T]CCCGCAGCTGCCGCTCCCCTTCCTG
- Phenotype
-
MIM: 157680
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDC25C
PubMed Links
Gene Details
- Gene
- CDC25C
- Gene Name
- cell division cycle 25C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001287582.1 |
1819 |
Missense Mutation |
GAG,GGG |
E462G |
NP_001274511.1 |
NM_001287583.1 |
1819 |
Missense Mutation |
GAG,GGG |
E540G |
NP_001274512.1 |
NM_001318098.1 |
1819 |
Missense Mutation |
GAG,GGG |
E479G |
NP_001305027.1 |
NM_001790.4 |
1819 |
Missense Mutation |
GAG,GGG |
E462G |
NP_001781.2 |
NM_022809.3 |
1819 |
Missense Mutation |
GAG,GGG |
E389G |
NP_073720.1 |
XM_005272145.3 |
1819 |
Missense Mutation |
GAG,GGG |
E506G |
XP_005272202.1 |
XM_006714739.3 |
1819 |
Missense Mutation |
GAG,GGG |
E464G |
XP_006714802.1 |
XM_011543759.2 |
1819 |
Missense Mutation |
GAG,GGG |
E486G |
XP_011542061.1 |
XM_011543760.2 |
1819 |
Missense Mutation |
GAG,GGG |
E485G |
XP_011542062.1 |
XM_011543761.2 |
1819 |
Missense Mutation |
GAG,GGG |
E467G |
XP_011542063.1 |
XM_011543762.1 |
1819 |
Missense Mutation |
GAG,GGG |
E389G |
XP_011542064.1 |
XM_011543763.1 |
1819 |
Missense Mutation |
GAG,GGG |
E351G |
XP_011542065.1 |
XM_011543764.1 |
1819 |
Missense Mutation |
GAG,GGG |
E300G |
XP_011542066.1 |
XM_017010101.1 |
1819 |
Missense Mutation |
GAG,GGG |
E428G |
XP_016865590.1 |
View Full Product Details