Product Details

SNP ID
rs78365495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:97160959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCAGTCTGGATAAATACACAAC[A/G]AAGGATCATAGCTGAAATACCAGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RIOK2 PubMed Links

Gene Details

Gene
RIOK2
Gene Name
RIO kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159749.1 3871 Intron NP_001153221.1
NM_018343.2 3871 UTR 3 NP_060813.2
XM_017009628.1 3871 UTR 3 XP_016865117.1

View Full Product Details