Product Details
- SNP ID
-
hCV105212496
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:145747316 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGGTAACTGCCCATTACTTCCCA[G/A]TATGGTCTAGAGCAGAGGTCAGCAA
- Phenotype
-
MIM: 604463
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD160
PubMed Links
Gene Details
- Gene
- CD160
- Gene Name
- CD160 molecule
There are no transcripts associated with this gene.
- Gene
- RNF115
- Gene Name
- ring finger protein 115
There are no transcripts associated with this gene.
View Full Product Details