Product Details

SNP ID
hCV105213830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:147158402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATGAGTTAAATGAACTTGAGCA[A/G]TTTATCTAGAGCAGTGTTTAGTGTA
Phenotype
MIM: 602741
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRKAB2 PubMed Links

Gene Details

Gene
PRKAB2
Gene Name
protein kinase AMP-activated non-catalytic subunit beta 2
There are no transcripts associated with this gene.

View Full Product Details