Product Details
- SNP ID
-
rs74756813
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
15
- Location
-
Chr.1:225985342 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCAAAGGCTTGGCAAATAAAGCC[A/G]GGTCAATTTGCTCCTTTGCCAGTCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SDE2
PubMed Links
Gene Details
- Gene
- SDE2
- Gene Name
- SDE2 telomere maintenance homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152608.3 |
1369 |
Missense Mutation |
CCG,CTG |
P439L |
NP_689821.3 |
View Full Product Details