Product Details

SNP ID
rs75017395
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:247815284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCATCACAGCAATCAGACCCCCA[C/T]ACAGCCAAGACACAGTGGCTCTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR14A16 PubMed Links

Gene Details

Gene
OR14A16
Gene Name
olfactory receptor family 14 subfamily A member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001966.1 446 Missense Mutation TAT,TGT Y149C NP_001001966.1

View Full Product Details