Product Details

SNP ID
rs74913817
Assay Type
Functionally Tested
NCBI dbSNP Submissions
13
Location
Chr.1:211390064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTCAGGAATGTGCCTAGGTTAT[A/G]TTTCTCTTGAAGTCCATGGTTTTTA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LINC00467 PubMed Links

Gene Details

Gene
LINC00467
Gene Name
long intergenic non-protein coding RNA 467
There are no transcripts associated with this gene.

View Full Product Details