Product Details

SNP ID
rs75063374
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:212797097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACTGTTCCTGCTTTCTCAGTTG[C/G]TTCTACATTTTTAGGTATAATGGGT
Phenotype
MIM: 609174
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NSL1 PubMed Links

Gene Details

Gene
NSL1
Gene Name
NSL1, MIS12 kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042549.1 Intron NP_001036014.1
NM_001297736.1 Intron NP_001284665.1
NM_001297737.1 Intron NP_001284666.1
NM_001297739.1 Intron NP_001284668.1
NM_015471.3 Intron NP_056286.3
XM_017000968.1 Intron XP_016856457.1
XM_017000969.1 Intron XP_016856458.1
Gene
TATDN3
Gene Name
TatD DNase domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042552.2 Intron NP_001036017.1
NM_001042553.2 Intron NP_001036018.1
NM_001146169.1 Intron NP_001139641.1
NM_001146170.1 Intron NP_001139642.1
NM_001146171.1 Intron NP_001139643.1
XM_011509202.2 Intron XP_011507504.1
XM_017000328.1 Intron XP_016855817.1
XM_017000329.1 Intron XP_016855818.1
XM_017000330.1 Intron XP_016855819.1
XM_017000331.1 Intron XP_016855820.1

View Full Product Details