Product Details

SNP ID
rs77486347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:25422157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTCTCAGAAAAGAGATAATTCC[C/T]TCCAATGATAATGAAATGATTTAAA
Phenotype
MIM: 616904 MIM: 152760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOCK5 PubMed Links

Gene Details

Gene
DOCK5
Gene Name
dedicator of cytokinesis 5
There are no transcripts associated with this gene.

Gene
GNRH1
Gene Name
gonadotropin releasing hormone 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000825.3 Intron NP_000816.4
NM_001083111.1 Intron NP_001076580.1
Gene
KCTD9
Gene Name
potassium channel tetramerization domain containing 9
There are no transcripts associated with this gene.

View Full Product Details